Please login to your online account to display your discounted pricing

2-Acetyl-5-methylthiophene 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Acetyl-5-methylthiophene
CAS 13679-74-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H8OS
Formula Weight 140.2
MDL Number MFCD00014529
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC382880050

  SDS
thermo fisher scientific
AC382880050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC382880250

  SDS
thermo fisher scientific
AC382880250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Low Melting Crystalline Mass, Powder or Crystals
Formula Weight 140.2
Quantity 5g
Color White to pale yellow
CAS Max % 100.0
Chemical Name or Material 2-Acetyl-5-methylthiophene
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
CAS 13679-74-8
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD00014529
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Molecular Formula C7H8OS
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.