missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Acetyl-5-methylthiophene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1128222
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Acetyl-5-methylthiophene | |
24°C to 28°C | |
65°C to 67°C (1 mmHg) | |
1.5625 | |
100 g | |
Light sensitive | |
YOSDTJYMDAEEAZ-UHFFFAOYSA-N | |
1-(5-methylthiophen-2-yl)ethanone | |
83655 | |
98% |
13679-74-8 | |
1.119 | |
C7H8OS | |
MFCD00014529 | |
110854 | |
2-acetyl-5-methylthiophene, 1-5-methylthiophen-2-yl ethanone, ketone, methyl 5-methyl-2-thienyl, 1-5-methyl-2-thienyl ethan-1-one, 2-methyl-5-acetylthiophene, ethanone, 1-5-methyl-2-thienyl, methylthienylcetone, 1-5-methyl-2-thienyl ethanone, 5-methyl-2-acetylthiophene, methyl 5-methyl-2-thienyl ketone | |
CC(=O)C1=CC=C(C)S1 | |
140.20 | |
140.2 |
Chemical Identifiers
13679-74-8 | |
140.20 | |
YOSDTJYMDAEEAZ-UHFFFAOYSA-N | |
83655 | |
CC(=O)C1=CC=C(C)S1 |
C7H8OS | |
MFCD00014529 | |
2-acetyl-5-methylthiophene, 1-5-methylthiophen-2-yl ethanone, ketone, methyl 5-methyl-2-thienyl, 1-5-methyl-2-thienyl ethan-1-one, 2-methyl-5-acetylthiophene, ethanone, 1-5-methyl-2-thienyl, methylthienylcetone, 1-5-methyl-2-thienyl ethanone, 5-methyl-2-acetylthiophene, methyl 5-methyl-2-thienyl ketone | |
1-(5-methylthiophen-2-yl)ethanone |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 237-181-2
RTECSNumber : OB4972000
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only