Please login to your online account to display your discounted pricing

2-Acetyl-5-chlorothiophene, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC306700250

 View more versions of this product

Catalog No. AC306700250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystals or Crystalline Powder
Formula Weight 160.62
Quantity 25g
Color White to light yellow
Melting Point 43° to 47°C
CAS Max % 100.0
Chemical Name or Material 2-Acetyl-5-chlorothiophene
Molecular Formula C6H5ClOS
CAS 6310-09-4
Packaging Glass bottle
Assay Percent Range 99%
MDL Number MFCD00005444
CAS Min % 98.5
Infrared Spectrum Authentic
Synonym 5-Chloro-2-thienyl methyl ketone

Specifications

Chemical Name or Material 2-Acetyl-5-chlorothiophene
CAS 6310-09-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H5ClOS
Formula Weight 160.62
MDL Number MFCD00005444
Synonym 5-Chloro-2-thienyl methyl ketone