Please login to your online account to display your discounted pricing

2-Acetyl-5-chlorothiophene, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Acetyl-5-chlorothiophene
CAS 6310-09-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H5ClOS
Formula Weight 160.62
MDL Number MFCD00005444
Synonym 5-Chloro-2-thienyl methyl ketone
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC306700250

  SDS
thermo fisher scientific
AC306700250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 160.62
Physical Form Crystals or Crystalline Powder
Quantity 25g
Color White to light yellow
Melting Point 43° to 47°C
CAS Max % 100.0
CAS 6310-09-4
Molecular Formula C6H5ClOS
Chemical Name or Material 2-Acetyl-5-chlorothiophene
CAS Min % 98.5
MDL Number MFCD00005444
Packaging Glass bottle
Assay Percent Range 99%
Infrared Spectrum Authentic
Synonym 5-Chloro-2-thienyl methyl ketone