missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-5-chlorothiophene, 99%

Manufacturer:  Alfa Aesar™ A1185018

 View more versions of this product

Catalog No. AAA1185018



Specifications

Specifications

2-Acetyl-5-chlorothiophene
117°C to 118°C (17mmHg)
50g
C6H5ClOS
UN2811
2-acetyl-5-chlorothiophene, 1-5-chlorothiophen-2-yl ethanone, 1-5-chloro-2-thienyl ethanone, 5-chloro-2-acetylthiophene, ethanone, 1-5-chloro-2-thienyl, 5-chlor-2-acetyl thiophen, ketone, 5-chloro-2-thienyl methyl, 1-5-chlorothiophen-2-yl ethan-1-one, 5-chloro-2-thienyl methyl ketone, 2-chloro-5-acetylthiophene
CC(=O)C1=CC=C(S1)Cl
160.62
160.62
108°C (226°F)
44°C to 48°C
6310-09-4
MFCD00005444
113936
HTZGPEHWQCRXGZ-UHFFFAOYSA-N
1-(5-chlorothiophen-2-yl)ethanone
80572
99%

Chemical Identifiers

6310-09-4
160.62
HTZGPEHWQCRXGZ-UHFFFAOYSA-N
80572
CC(=O)C1=CC=C(S1)Cl
C6H5ClOS
MFCD00005444
2-acetyl-5-chlorothiophene, 1-5-chlorothiophen-2-yl ethanone, 1-5-chloro-2-thienyl ethanone, 5-chloro-2-acetylthiophene, ethanone, 1-5-chloro-2-thienyl, 5-chlor-2-acetyl thiophen, ketone, 5-chloro-2-thienyl methyl, 1-5-chlorothiophen-2-yl ethan-1-one, 5-chloro-2-thienyl methyl ketone, 2-chloro-5-acetylthiophene
1-(5-chlorothiophen-2-yl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H301-H312-H332
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : OB1745000

SDS Documents