Learn More
2-Acetyl-5-bromothiophene, 99%, Thermo Scientific Chemicals
$74.16 - $1,026.11
Chemical Identifiers
CAS | 5370-25-2 |
---|---|
Molecular Formula | C6H5BrOS |
Molecular Weight (g/mol) | 205.069 |
MDL Number | MFCD00014528 |
InChI Key | IGBZCOWXSCWSHO-UHFFFAOYSA-N |
Synonym | 2-acetyl-5-bromothiophene, 1-5-bromothiophen-2-yl ethanone, ethanone, 1-5-bromo-2-thienyl, 1-5-bromo-2-thienyl ethan-1-one, 1-5-bromothiophen-2-yl ethan-1-one, 5-bromo-2-thienyl methyl ketone, 2-acetyl-5-bromo thiophene, ketone, 5-bromo-2-thienyl methyl, 2-acetyl-5-bromo-thiophene, 1-5-bromo-2-thienyl ethanone |
PubChem CID | 79335 |
IUPAC Name | 1-(5-bromothiophen-2-yl)ethanone |
SMILES | CC(=O)C1=CC=C(S1)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1482609
|
Thermo Scientific Chemicals
A1482609 |
10 g |
N/A
|
|
|||||
AAA1482618
|
Thermo Scientific Chemicals
A1482618 |
50 g |
N/A
|
|
|||||
AAA1482630
|
Thermo Scientific Chemicals
A1482630 |
250 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5370-25-2 | |
205.069 | |
IGBZCOWXSCWSHO-UHFFFAOYSA-N | |
79335 | |
CC(=O)C1=CC=C(S1)Br |
C6H5BrOS | |
MFCD00014528 | |
2-acetyl-5-bromothiophene, 1-5-bromothiophen-2-yl ethanone, ethanone, 1-5-bromo-2-thienyl, 1-5-bromo-2-thienyl ethan-1-one, 1-5-bromothiophen-2-yl ethan-1-one, 5-bromo-2-thienyl methyl ketone, 2-acetyl-5-bromo thiophene, ketone, 5-bromo-2-thienyl methyl, 2-acetyl-5-bromo-thiophene, 1-5-bromo-2-thienyl ethanone | |
1-(5-bromothiophen-2-yl)ethanone |
Specifications
5370-25-2 | |
103°C (4 mmHg) | |
MFCD00014528 | |
113949 | |
2-acetyl-5-bromothiophene, 1-5-bromothiophen-2-yl ethanone, ethanone, 1-5-bromo-2-thienyl, 1-5-bromo-2-thienyl ethan-1-one, 1-5-bromothiophen-2-yl ethan-1-one, 5-bromo-2-thienyl methyl ketone, 2-acetyl-5-bromo thiophene, ketone, 5-bromo-2-thienyl methyl, 2-acetyl-5-bromo-thiophene, 1-5-bromo-2-thienyl ethanone | |
CC(=O)C1=CC=C(S1)Br | |
205.069 | |
205.07 | |
2-Acetyl-5-bromothiophene |
93°C to 96°C | |
C6H5BrOS | |
10 g | |
Light sensitive | |
IGBZCOWXSCWSHO-UHFFFAOYSA-N | |
1-(5-bromothiophen-2-yl)ethanone | |
79335 | |
99% |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 226-363-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only