Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-5-bromobenzo[b]furan, 99%

$545.45 - $545.45

Chemical Identifiers

Chemical Name or Material 2-Acetyl-5-bromobenzo[b]furan
CAS 38220-75-6
Molecular Formula C10H7BrO2
Formula Weight 239.07
MDL Number MFCD00276737
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5059306

  SDS
alfa aesar™
H5059306
5g Each for $545.45

AAH5059303

  SDS
alfa aesar™
H5059303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Acetyl-5-bromobenzo[b]furan
CAS 38220-75-6
Assay 99%
Molecular Formula C10H7BrO2
Formula Weight 239.07
MDL Number MFCD00276737
Quantity 5g