missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-4-methylthiophene, 98%

$33.61 - $73.57

Chemical Identifiers

CAS 13679-73-7
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00027395
InChI Key NFZNHUNPIJPRMP-UHFFFAOYSA-N
Synonym 2-acetyl-4-methylthiophene, 1-4-methylthiophen-2-yl ethanone, 1-4-methyl-2-thienyl ethanone, ethanone, 1-4-methyl-2-thienyl, 1-4-methyl-2-thienyl ethan-1-one, 1-4-methyl-thiophen-2-yl-ethanone, 2-acetyl-4-methylthiophene, 4-methyl-2-acetylthiophene, pubchem5214, acmc-20aa2e
PubChem CID 83654
IUPAC Name 1-(4-methylthiophen-2-yl)ethanone
SMILES CC1=CSC(=C1)C(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1761703 View Documents Alfa Aesar™
L1761703
1g Each for $33.61
 
AAL1761706 View Documents Alfa Aesar™
L1761706
5g Each for $73.57
 
Specifications

Chemical Identifiers

13679-73-7
140.2
NFZNHUNPIJPRMP-UHFFFAOYSA-N
83654
CC1=CSC(=C1)C(=O)C
C7H8OS
MFCD00027395
2-acetyl-4-methylthiophene, 1-4-methylthiophen-2-yl ethanone, 1-4-methyl-2-thienyl ethanone, ethanone, 1-4-methyl-2-thienyl, 1-4-methyl-2-thienyl ethan-1-one, 1-4-methyl-thiophen-2-yl-ethanone, 2-acetyl-4-methylthiophene, 4-methyl-2-acetylthiophene, pubchem5214, acmc-20aa2e
1-(4-methylthiophen-2-yl)ethanone

Specifications

97°C to 99°C (8mmHg)
13679-73-7
MFCD00027395
2-acetyl-4-methylthiophene, 1-4-methylthiophen-2-yl ethanone, 1-4-methyl-2-thienyl ethanone, ethanone, 1-4-methyl-2-thienyl, 1-4-methyl-2-thienyl ethan-1-one, 1-4-methyl-thiophen-2-yl-ethanone, 2-acetyl-4-methylthiophene, 4-methyl-2-acetylthiophene, pubchem5214, acmc-20aa2e
CC1=CSC(=C1)C(=O)C
140.2
140.2
2-Acetyl-4-methylthiophene
1g
C7H8OS
111378
NFZNHUNPIJPRMP-UHFFFAOYSA-N
1-(4-methylthiophen-2-yl)ethanone
83654
98%
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger