Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-4-fluorobenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 2-Acetyl-4-fluorobenzeneboronic acid pinacol ester
Formula Weight 264.1
Synonym 2-Acetyl-4-fluorophenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6242703

 
alfa aesar™
H6242703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62427MD

 
alfa aesar™
H62427MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 264.1
Chemical Name or Material 2-Acetyl-4-fluorobenzeneboronic acid pinacol ester
Physical Form Solid
Assay 96%
Synonym 2-Acetyl-4-fluorophenylboronic acid pinacol ester
Quantity 1g