Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-4-bromopyridine, 98%

Chemical Identifiers

Chemical Name or Material 2-Acetyl-4-bromopyridine
CAS 1060805-69-7
Formula Weight 200.04
MDL Number MFCD13189212
Beilstein 22133194
Synonym 1-(4-Bromo-2-pyridyl)ethanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH66562MD

 
alfa aesar™
H66562MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6656203

 
alfa aesar™
H6656203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.04
Chemical Name or Material 2-Acetyl-4-bromopyridine
CAS 1060805-69-7
Physical Form Solid
MDL Number MFCD13189212
Beilstein 22133194
Synonym 1-(4-Bromo-2-pyridyl)ethanone
Assay 98%
Quantity 250mg