Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole, 97%

Chemical Identifiers

Chemical Name or Material 2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole
CAS 94944-70-4
Formula Weight 230.22
MDL Number MFCD08752516
Synonym 1-[4-(1,2,3,4-Tetrahydroxybutyl)imidazol-2-yl]ethanone; THI
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH63416MD

 
alfa aesar™
H63416MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 230.22
Chemical Name or Material 2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole
Physical Form Powder
Assay 97%
MDL Number MFCD08752516
CAS 94944-70-4
Synonym 1-[4-(1,2,3,4-Tetrahydroxybutyl)imidazol-2-yl]ethanone; THI
Health Hazard 1 Warning
Color Off-white to pale brown
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Quantity 250mg
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C

Applications
2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole used for treatment of autoimmune disorders.

Solubility
Soluble in distilled water, DMSO (>5 mg/ml), clear (warmed), and PBS pH 7.2 (~0.15 mg/ml).

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.

2° to 8°C