Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-3-methylthiophene, 97%

$56.50 - $195.88

Chemical Identifiers

Chemical Name or Material 2-Acetyl-3-methylthiophene
CAS 13679-72-6
Molecular Formula C7H8OS
Formula Weight 140.2
MDL Number MFCD00005443
Beilstein 112023
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0408603

  SDS
alfa aesar™
L0408603
1g Each for $56.50

AAL0408606

  SDS
alfa aesar™
L0408606
5g Each for $195.88
Description & Specifications

Specifications

Formula Weight 140.2
Chemical Name or Material 2-Acetyl-3-methylthiophene
CAS 13679-72-6
Molecular Formula C7H8OS
MDL Number MFCD00005443
Health Hazard 1 Danger
Assay 97%
Flash Point 92°C (197°F)
Boiling Point 68° to 72°C (3mmHg)
Health Hazard 2 GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P405-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 112023
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P280-P301+P310-P405-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.