missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-3-methylpyrazine, 98%

Manufacturer:  Alfa Aesar™ A1505414

 View more versions of this product

Catalog No. AAA1505414



Specifications

Specifications

2-Acetyl-3-methylpyrazine
265°C to 266°C
23787-80-6
MFCD00014612
2-acetyl-3-methylpyrazine, 1-3-methylpyrazin-2-yl ethanone, 1-3-methylpyrazin-2-yl ethan-1-one, ethanone, 1-3-methylpyrazinyl, 2-methyl-3-acetylpyrazine, unii-7u5tew2y2b, 2-acetyl-3-methyl pyrazine, 1-3-methylpyrazinyl ethan-1-one, 3-acetyl-2-methylpyrazine, ethanone, 1-3-methyl-2-pyrazinyl
CC1=NC=CN=C1C(=O)C
136.154
136.15
>100°C (212°F)
25g
C7H8N2O
742438
QUNOTZOHYZZWKQ-UHFFFAOYSA-N
1-(3-methylpyrazin-2-yl)ethanone
32093
98%

Chemical Identifiers

23787-80-6
136.154
QUNOTZOHYZZWKQ-UHFFFAOYSA-N
32093
CC1=NC=CN=C1C(=O)C
C7H8N2O
MFCD00014612
2-acetyl-3-methylpyrazine, 1-3-methylpyrazin-2-yl ethanone, 1-3-methylpyrazin-2-yl ethan-1-one, ethanone, 1-3-methylpyrazinyl, 2-methyl-3-acetylpyrazine, unii-7u5tew2y2b, 2-acetyl-3-methyl pyrazine, 1-3-methylpyrazinyl ethan-1-one, 3-acetyl-2-methylpyrazine, ethanone, 1-3-methyl-2-pyrazinyl
1-(3-methylpyrazin-2-yl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents