Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-3-methylpyrazine, 98%

$69.46 - $971.86

Chemical Identifiers

Chemical Name or Material 2-Acetyl-3-methylpyrazine
CAS 23787-80-6
Molecular Formula C7H8N2O
Formula Weight 136.15
MDL Number MFCD00014612
Beilstein 742438
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1505403

  SDS
alfa aesar™
A1505403
1g Each for $69.46

AAA1505406

  SDS
alfa aesar™
A1505406
5g Each for $235.05

AAA1505414

  SDS
alfa aesar™
A1505414
25g Each for $971.86
Description & Specifications

Specifications

Formula Weight 136.15
Beilstein 742438
Flash Point >100°C (212°F)
Chemical Name or Material 2-Acetyl-3-methylpyrazine
Assay 98%
CAS 23787-80-6
Health Hazard 1 Warning
Molecular Formula C7H8N2O
Quantity 1g
TSCA TSCA
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 265° to 266°C
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
MDL Number MFCD00014612
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.