Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-3-methylbenzo[b]thiophene, 97%

$58.04 - $711.15

Chemical Identifiers

Chemical Name or Material 2-Acetyl-3-methylbenzo[b]thiophene
CAS 18781-31-2
Molecular Formula C11H10OS
Formula Weight 190.27
MDL Number MFCD00052499
Beilstein 124400
Synonym 2-Acetyl-3-methylthianaphthene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2096702

 
alfa aesar™
B2096702
0.5g Each for $58.04

AAB2096709

 
alfa aesar™
B2096709
10g Each for $711.15
Description & Specifications

Specifications

Formula Weight 190.27
Chemical Name or Material 2-Acetyl-3-methylbenzo[b]thiophene
Assay 97%
Molecular Formula C11H10OS
MDL Number MFCD00052499
CAS 18781-31-2
Beilstein 124400
Synonym 2-Acetyl-3-methylthianaphthene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 76° to 80°C
Boiling Point 129° to 131°C (0.5mmHg)
Quantity 0.5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.