Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-3-ethylpyrazine, 98%

$43.84 - $170.27

Chemical Identifiers

Chemical Name or Material 2-Acetyl-3-ethylpyrazine
CAS 32974-92-8
Molecular Formula C8H10N2O
Formula Weight 150.18
MDL Number MFCD00038028
Beilstein 742901
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1567403

  SDS
alfa aesar™
A1567403
1g Each for $43.84

AAA1567406

  SDS
alfa aesar™
A1567406
5g Each for $170.27
Description & Specifications

Specifications

Formula Weight 150.18
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 32974-92-8
TSCA TSCA
Beilstein 742901
Boiling Point 54° to 56°C (1mmHg)
Molecular Formula C8H10N2O
Flash Point 87°C (188°F)
Health Hazard 1 Warning
Assay 98%
MDL Number MFCD00038028
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Acetyl-3-ethylpyrazine
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.