missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-3-ethylpyrazine, 98%

Manufacturer:  Alfa Aesar™ A1567403

 View more versions of this product

Catalog No. AAA1567403



Specifications

Specifications

2-Acetyl-3-ethylpyrazine
54°C to 56°C (1mmHg)
32974-92-8
MFCD00038028
2-acetyl-3-ethylpyrazine, 1-3-ethylpyrazin-2-yl ethanone, ethanone, 1-3-ethylpyrazinyl, 2-acetyl-3-ethyl-1,4-diazine, 1-3-ethylpyrazinyl ethan-1-one, unii-aq03n14xtl, 1-3-ethylpyrazinyl ethanone, 1-3-ethyl-2-pyrazinyl ethanone, 3-ethyl-2-acetylpyrazine, 2-acetyl-3-ethyl pyrazine
CCC1=NC=CN=C1C(=O)C
150.181
150.18
87°C (188°F)
1g
C8H10N2O
742901
PPJSYGVFDJEMRP-UHFFFAOYSA-N
1-(3-ethylpyrazin-2-yl)ethanone
61918
98%

Chemical Identifiers

32974-92-8
150.181
PPJSYGVFDJEMRP-UHFFFAOYSA-N
61918
CCC1=NC=CN=C1C(=O)C
C8H10N2O
MFCD00038028
2-acetyl-3-ethylpyrazine, 1-3-ethylpyrazin-2-yl ethanone, ethanone, 1-3-ethylpyrazinyl, 2-acetyl-3-ethyl-1,4-diazine, 1-3-ethylpyrazinyl ethan-1-one, unii-aq03n14xtl, 1-3-ethylpyrazinyl ethanone, 1-3-ethyl-2-pyrazinyl ethanone, 3-ethyl-2-acetylpyrazine, 2-acetyl-3-ethyl pyrazine
1-(3-ethylpyrazin-2-yl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents