Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-3-chlorothiophene, 97%

Chemical Identifiers

Chemical Name or Material 2-Acetyl-3-chlorothiophene
CAS 89581-82-8
Molecular Formula C6H5ClOS
Formula Weight 160.62
MDL Number MFCD00794248
UN Number UN2810
Beilstein 10307002
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2011403

  SDS
alfa aesar™
L2011403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2011406

  SDS
alfa aesar™
L2011406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2011414

  SDS
alfa aesar™
L2011414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 160.62
Assay 97%
CAS 89581-82-8
Chemical Name or Material 2-Acetyl-3-chlorothiophene
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00794248
Molecular Formula C6H5ClOS
Quantity 1g
Beilstein 10307002
UN Number UN2810
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Boiling Point 117° to 118°C (15mmHg)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H312-H332
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H312-H332
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.