Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-3-bromothiophene, 97%

$39.57 - $263.22

Chemical Identifiers

Chemical Name or Material 2-Acetyl-3-bromothiophene
CAS 42877-08-7
Molecular Formula C6H5BrOS
Formula Weight 205.07
MDL Number MFCD00794287
Beilstein 116973
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2011303

 
alfa aesar™
L2011303
1g Each for $39.57

AAL2011306

 
alfa aesar™
L2011306
5g Each for $69.58

AAL2011314

 
alfa aesar™
L2011314
25g Each for $263.22
Description & Specifications

Specifications

Formula Weight 205.07
Molecular Formula C6H5BrOS
Chemical Name or Material 2-Acetyl-3-bromothiophene
MDL Number MFCD00794287
CAS 42877-08-7
Beilstein 116973
Health Hazard 1 Warning
Melting Point 41° to 45°C
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Boiling Point 102° to 105°C (4mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.