Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-3-amino-5-phenylthiophene, 97%

$103.35 - $400.80

Chemical Identifiers

Chemical Name or Material 2-Acetyl-3-amino-5-phenylthiophene
CAS 105707-24-2
Molecular Formula C12H11NOS
Formula Weight 217.29
MDL Number MFCD00068132
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2075102

  SDS
alfa aesar™
B2075102
0.5g Each for $103.35

AAB2075188

  SDS
alfa aesar™
B2075188
2.5g Each for $400.80
Description & Specifications

Specifications

Formula Weight 217.29
Health Hazard 1 Warning
MDL Number MFCD00068132
Assay 97%
Quantity 0.5g
Molecular Formula C12H11NOS
CAS 105707-24-2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Acetyl-3-amino-5-phenylthiophene
Melting Point 119° to 122°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.