Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-3,6-difluorobenzoic acid, 98%

$710.24 - $710.24

Chemical Identifiers

Chemical Name or Material 2-Acetyl-3,6-difluorobenzoic acid
CAS 319457-34-6
Molecular Formula C9H6F2O3
Formula Weight 200.14g/mol
MDL Number MFCD00800682
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2610003

 
alfa aesar™
H26100-03
1g Each for $710.24
Description & Specifications

Specifications

Melting Point 83° to 88°C
Quantity 1g
Chemical Name or Material 2-Acetyl-3,6-difluorobenzoic acid
CAS 319457-34-6
Assay 98%
Molecular Formula C9H6F2O3
Formula Weight 200.14g/mol
MDL Number MFCD00800682
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.