Please login to your online account to display your discounted pricing

2-Acetyl-1-naphthol, 99%, ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  195000250

 View more versions of this product

Catalog No. AC195000250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 186.21
Physical Form Crystalline Powder
Quantity 25g
Color Yellow
Beilstein 08, 149
TSCA TSCA
EINECS Number 211-918-8
Melting Point 97° to 100°C
CAS Max % 100.0
Synonym 1'-Hydroxy-2'-acetonaphthone
Health Hazard 1 GHS Signal Word: Warning
Packaging Plastic bottle
Infrared Spectrum Authentic
Molecular Formula C12H10O2
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99%
Chemical Name or Material 2-Acetyl-1-naphthol
CAS 711-79-5
CAS Min % 98.5
Assay Percent Range 99%
Linear Formula HOC10H6COCH3
MDL Number MFCD00003963
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.

Specifications

Chemical Name or Material 2-Acetyl-1-naphthol
Name Note 99%
CAS 711-79-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C12H10O2
Linear Formula HOC10H6COCH3
Formula Weight 186.21
MDL Number MFCD00003963
Synonym 1'-Hydroxy-2'-acetonaphthone
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.

SDS & Safety Documents