missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-1-methylpyrrole, 98%

$60.66 - $694.22

Chemical Identifiers

CAS 932-16-1
Molecular Formula C7H9NO
Formula Weight 123.16
MDL Number MFCD00003089
Beilstein 111887
Chemical Name or Material 2-Acetyl-1-methylpyrrole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1535306 SDS Alfa Aesar™
A1535306
5g Each for $60.66
AAA1535314 SDS Alfa Aesar™
A1535314
25g Each for $218.58
AAA1535322 SDS Alfa Aesar™
A1535322
100g Each for $694.22
Specifications Description & Specifications

Chemical Identifiers

932-16-1
123.16
111887
C7H9NO
MFCD00003089
2-Acetyl-1-methylpyrrole

Specifications

932-16-1
123.16
111887
5g
Pleasant
98%
C7H9NO
MFCD00003089
200° to 202°C
68°C (154°F)
2-Acetyl-1-methylpyrrole

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA