missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-1-methylpyrrole, 98%

Manufacturer:  Alfa Aesar™ A1535322

 View more versions of this product

Catalog No. AAA1535322



Specifications

Specifications

2-Acetyl-1-methylpyrrole
Pleasant
100g
C7H9NO
111887
NZFLWVDXYUGFAV-UHFFFAOYSA-N
1-(1-methylpyrrol-2-yl)ethanone
61240
123.16
68°C (154°F)
200°C to 202°C
932-16-1
MFCD00003089
2-acetyl-1-methylpyrrole, n-methyl-2-acetylpyrrole, 1-1-methyl-1h-pyrrol-2-yl ethanone, 1-1-methylpyrrol-2-yl ethanone, 1-methyl-2-acetylpyrrole, 2-acetyl-n-methylpyrrole, ethanone, 1-1-methyl-1h-pyrrol-2-yl, methyl 1-methylpyrrol-2-yl ketone, 1-1-methyl-1h-pyrrol-2-yl ethan-1-one, 2-acetyl-1-methyl pyrrole
CC(=O)C1=CC=CN1C
123.155
CHEBI:59982
98%

Chemical Identifiers

932-16-1
123.155
NZFLWVDXYUGFAV-UHFFFAOYSA-N
61240
1-(1-methylpyrrol-2-yl)ethanone
C7H9NO
MFCD00003089
2-acetyl-1-methylpyrrole, n-methyl-2-acetylpyrrole, 1-1-methyl-1h-pyrrol-2-yl ethanone, 1-1-methylpyrrol-2-yl ethanone, 1-methyl-2-acetylpyrrole, 2-acetyl-n-methylpyrrole, ethanone, 1-1-methyl-1h-pyrrol-2-yl, methyl 1-methylpyrrol-2-yl ketone, 1-1-methyl-1h-pyrrol-2-yl ethan-1-one, 2-acetyl-1-methyl pyrrole
CHEBI:59982
CC(=O)C1=CC=CN1C
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents