Please login to your online account to display your discounted pricing

2-Acetoxyphenylboronic acid pinacol ester, 97%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC444420010

 View more versions of this product

Catalog No. AC444420010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 262.12
Quantity 1g
Molecular Formula C14H19BO4
Synonym 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate
Chemical Name or Material 2-Acetoxyphenylboronic acid pinacol ester
Assay Percent Range 97%

Specifications

Chemical Name or Material 2-Acetoxyphenylboronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C14H19BO4
Formula Weight 262.12
Synonym 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate