Please login to your online account to display your discounted pricing

2-Acetoxyphenylboronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Acetoxyphenylboronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C14H19BO4
Formula Weight 262.12
Synonym 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC444420010

 
thermo fisher scientific
AC444420010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC444420050

 
thermo fisher scientific
AC444420050
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 262.12
Quantity 1g
Chemical Name or Material 2-Acetoxyphenylboronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C14H19BO4
Synonym 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate