Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetoxyisobutyryl chloride, 98%

Chemical Identifiers

Chemical Name or Material 2-Acetoxyisobutyryl chloride
CAS 40635-66-3
Molecular Formula C6H9ClO3
Formula Weight 164.59
MDL Number MFCD00000708
UN Number UN3265
Beilstein 507772
Synonym 2-Acetoxy-2-methylpropionyl chloride; 1-Chlorocarbonyl-1-methylethyl acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0095709

  SDS
alfa aesar™
L0095709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0095718

  SDS
alfa aesar™
L0095718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 164.59
Chemical Name or Material 2-Acetoxyisobutyryl chloride
Molecular Formula C6H9ClO3
MDL Number MFCD00000708
Synonym 2-Acetoxy-2-methylpropionyl chloride; 1-Chlorocarbonyl-1-methylethyl acetate
Boiling Point 55° to 56°C (6mmHg)
UN Number UN3265
Assay 98%
Health Hazard 2 GHS H Statement
H314-H318-H335-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Combustible liquid.
CAS 40635-66-3
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Beilstein 507772
Quantity 10g
Flash Point 68°C (154°F)
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H335-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Combustible liquid.