missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetoxyisobutyryl bromide, 96%

Manufacturer:  Alfa Aesar™ L1159206

 View more versions of this product

Catalog No. AAL1159206



Specifications

Specifications

2-Acetoxyisobutyryl bromide
100°C to 110°C (25mmHg)
40635-67-4
MFCD00040923
2432585
OOKAXSHFTDPZHP-UHFFFAOYSA-N
(1-bromo-2-methyl-1-oxopropan-2-yl) acetate
2724715
96%
>110°C (230°F)
5g
C6H9BrO3
UN3265
2-acetoxyisobutyryl bromide, 2-acetoxy-2-methylpropionyl bromide, 1-bromocarbonyl-1-methylethyl acetate, alpha-acetoxy-isobutyryl bromide, a-acetoxyisobutyryl bromide, alpha-acetoxyisobutyryl bromide, 2-acetoxyisobutyryl bromide;2-acetoxy-2-methylpropionyl bromide, propanoyl bromide, 2-acetyloxy-2-methyl, aibb, pubchem17437
CC(=O)OC(C)(C)C(=O)Br
209.039
209.05

Chemical Identifiers

40635-67-4
209.039
OOKAXSHFTDPZHP-UHFFFAOYSA-N
2724715
CC(=O)OC(C)(C)C(=O)Br
C6H9BrO3
MFCD00040923
2-acetoxyisobutyryl bromide, 2-acetoxy-2-methylpropionyl bromide, 1-bromocarbonyl-1-methylethyl acetate, alpha-acetoxy-isobutyryl bromide, a-acetoxyisobutyryl bromide, alpha-acetoxyisobutyryl bromide, 2-acetoxyisobutyryl bromide;2-acetoxy-2-methylpropionyl bromide, propanoyl bromide, 2-acetyloxy-2-methyl, aibb, pubchem17437
(1-bromo-2-methyl-1-oxopropan-2-yl) acetate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents