Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetoxycinnamic acid, predominantly trans, 98+%

$41.30 - $41.30

Chemical Identifiers

Chemical Name or Material 2-Acetoxycinnamic acid
CAS 55620-18-3
Name Note predominantly trans
Molecular Formula C11H10O4
Formula Weight 206.2
MDL Number MFCD00014375
Beilstein 2694007
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0284903

  SDS
alfa aesar™
L0284903
1g Each for $41.30
Description & Specifications

Specifications

Formula Weight 206.2
Chemical Name or Material 2-Acetoxycinnamic acid
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
MDL Number MFCD00014375
Assay ≥98%
Health Hazard 1 Warning
Name Note predominantly trans
CAS 55620-18-3
Quantity 1g
Molecular Formula C11H10O4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 2694007
Melting Point 154° to 158°C
RTECS Number GD7875000
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.