Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Acetoxyacetophenone, 98%

$38.18 - $455.62

Chemical Identifiers

Chemical Name or Material 2'-Acetoxyacetophenone
CAS 7250-94-4
Molecular Formula C10H10O3
Formula Weight 178.19
MDL Number MFCD00017223
Beilstein 1948938
Synonym 2-Acetylphenyl acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2162406

 
alfa aesar™
B2162406
5g Each for $38.18

AAB2162422

 
alfa aesar™
B2162422
100g Each for $455.62
Description & Specifications

Specifications

Formula Weight 178.19
Chemical Name or Material 2'-Acetoxyacetophenone
Assay 98%
Molecular Formula C10H10O3
MDL Number MFCD00017223
CAS 7250-94-4
Beilstein 1948938
Synonym 2-Acetylphenyl acetate
Boiling Point 118°C (19mmHg)
Melting Point 88° to 91°C
Quantity 5g