Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetoxyacetophenone, 98%

$79.58 - $218.58

Chemical Identifiers

Chemical Name or Material 2-Acetoxyacetophenone
CAS 2243-35-8
Molecular Formula C10H10O3
Formula Weight 178.19
MDL Number MFCD00026205
Beilstein 1240545
Synonym Acetic acid phenacyl ester; Phenacyl acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1565214

  SDS
alfa aesar™
L1565214
25g Each for $79.58

AAL1565222

  SDS
alfa aesar™
L1565222
100g Each for $218.58
Description & Specifications

Specifications

Formula Weight 178.19
Chemical Name or Material 2-Acetoxyacetophenone
TSCA TSCA
Assay 98%
MDL Number MFCD00026205
CAS 2243-35-8
Beilstein 1240545
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 110° to 114°C (2mmHg)
Melting Point 48° to 50°C
Flash Point >110°C (230°F)
Molecular Formula C10H10O3
Synonym Acetic acid phenacyl ester; Phenacyl acetate
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.