missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Acetonaphthone, 99%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 158885000

 View more versions of this product

Catalog No. AC158885000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2'-Acetonaphthone
Authentic
300.0°C to 301.0°C
53.0°C to 56.0°C
99%
98.5
98.5% min. (GC)
C10H7COCH3
07, 402
Solubility in water: insoluble
CC(=O)C1=CC2=CC=CC=C2C=C1
170.21
CHEBI:52364
Fine Crystalline Powder and Chunks
168°C
Plastic bottle
White
500g
93-08-3
100.0
C12H10O
MFCD00004108
1-2-naphthyl ethanone
XSAYZAUNJMRRIR-UHFFFAOYSA-N
1-naphthalen-2-ylethanone
7122
170.21
99%

Chemical Identifiers

93-08-3
170.21
XSAYZAUNJMRRIR-UHFFFAOYSA-N
7122
1-naphthalen-2-ylethanone
C12H10O
MFCD00004108
1-2-naphthyl ethanone
CHEBI:52364
CC(=O)C1=CC2=CC=CC=C2C=C1
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Warning

EINECSNumber : 202-216-2

RTECSNumber : AL2988000

TSCA : TSCA

SDS Documents