missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Acetamidothiazole, 98%, Thermo Scientific™
$185.88 - $185.88
Chemical Identifiers
CAS | 2719-23-5 |
---|---|
Molecular Formula | C5H6N2OS |
Molecular Weight (g/mol) | 142.176 |
MDL Number | MFCD00016892 |
InChI Key | WXPLRSVMGRAIGW-UHFFFAOYSA-N |
Synonym | 2-acetamidothiazole, 2-acetylaminothiazole, n-thiazol-2-yl acetamide, acetamide, n-2-thiazolyl, n-1,3-thiazol-2-yl acetamide, n-2-thiazolylacetamide, thiazole, 2-acetamido, 2-acetaminothiazole, 2-acetamido-1,3-thiazole, n-thiazol-2-ylacetamide |
PubChem CID | 17625 |
IUPAC Name | N-(1,3-thiazol-2-yl)acetamide |
SMILES | CC(=O)NC1=NC=CS1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2387314
|
Thermo Scientific Chemicals
B2387314 |
25 g |
Each for $185.88
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
2719-23-5 | |
142.176 | |
WXPLRSVMGRAIGW-UHFFFAOYSA-N | |
17625 | |
CC(=O)NC1=NC=CS1 |
C5H6N2OS | |
MFCD00016892 | |
2-acetamidothiazole, 2-acetylaminothiazole, n-thiazol-2-yl acetamide, acetamide, n-2-thiazolyl, n-1,3-thiazol-2-yl acetamide, n-2-thiazolylacetamide, thiazole, 2-acetamido, 2-acetaminothiazole, 2-acetamido-1,3-thiazole, n-thiazol-2-ylacetamide | |
N-(1,3-thiazol-2-yl)acetamide |
Specifications
2719-23-5 | |
C5H6N2OS | |
25 g | |
WXPLRSVMGRAIGW-UHFFFAOYSA-N | |
N-(1,3-thiazol-2-yl)acetamide | |
17625 | |
98% |
202°C to 204°C | |
MFCD00016892 | |
2-acetamidothiazole, 2-acetylaminothiazole, n-thiazol-2-yl acetamide, acetamide, n-2-thiazolyl, n-1,3-thiazol-2-yl acetamide, n-2-thiazolylacetamide, thiazole, 2-acetamido, 2-acetaminothiazole, 2-acetamido-1,3-thiazole, n-thiazol-2-ylacetamide | |
CC(=O)NC1=NC=CS1 | |
142.176 | |
142.18 | |
2-Acetamidothiazole |
Safety and Handling
EINECSNumber : 220-321-1
RTECSNumber : AC8800000
TSCA : Yes
Recommended Storage : Ambient temperatures