missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Acetamidophenol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2126414

 View more versions of this product

Catalog No. AAB2126414



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Acetamidophenol
205°C to 210°C
MFCD00002181
607592
ADVGKWPZRIDURE-UHFFFAOYSA-N
N-(2-hydroxyphenyl)acetamide
11972
97%
614-80-2
C8H9NO2
25 g
2-acetamidophenol, n-2-hydroxyphenyl acetamide, 2'-hydroxyacetanilide, o-hydroxyacetanilide, 2-acetaminophenol, 2-hydroxyacetanilide, o-acetamidophenol, o-acetylamino phenol, 2-acetylamino phenol, o-acetaminophenol
CC(=O)NC1=CC=CC=C1O
151.165
151.17

Chemical Identifiers

614-80-2
151.165
ADVGKWPZRIDURE-UHFFFAOYSA-N
11972
CC(=O)NC1=CC=CC=C1O
C8H9NO2
MFCD00002181
2-acetamidophenol, n-2-hydroxyphenyl acetamide, 2'-hydroxyacetanilide, o-hydroxyacetanilide, 2-acetaminophenol, 2-hydroxyacetanilide, o-acetamidophenol, o-acetylamino phenol, 2-acetylamino phenol, o-acetaminophenol
N-(2-hydroxyphenyl)acetamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H302-H315-H319

EINECSNumber : 210-396-9

RTECSNumber : AE4025000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only