Please login to your online account to display your discounted pricing

2-Acetamidofluorene, 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC102300250

 View more versions of this product

Catalog No. AC102300250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 223.27
Quantity 25g
Color White to yellow-brown
Melting Point 192° to 196°C
Beilstein 12, 1331
Merck Index 15, 4189
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 2-Acetamidofluorene
Packaging Glass bottle
Assay Percent Range 95%
Molecular Formula C15H13NO
MDL Number MFCD00001116
CAS 53-96-3
Infrared Spectrum Authentic
Synonym N-(2-Fluorenyl)acetamide
Health Hazard 2 GHS H Statement:
May cause cancer.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Acetamidofluorene
CAS 53-96-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C15H13NO
Formula Weight 223.27
MDL Number MFCD00001116
Synonym N-(2-Fluorenyl)acetamide
Accessories

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause cancer.
Harmful if swallowed.

SDS & Safety Documents