Please login to your online account to display your discounted pricing

2-Acetamidofluorene, 95%, ACROS Organics™

$152.14 - $152.14

Chemical Identifiers

Chemical Name or Material 2-Acetamidofluorene
CAS 53-96-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C15H13NO
Formula Weight 223.27
MDL Number MFCD00001116
Synonym N-(2-Fluorenyl)acetamide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC102300250

  SDS
thermo fisher scientific
AC102300250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC102300050

  SDS
thermo fisher scientific
102300050
Glass bottle 5g Each for $152.14
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 223.27
Physical Form Crystalline Powder
Quantity 25g
Color White to yellow-brown
Melting Point 192° to 196°C
Beilstein 12, 1331
Merck Index 15, 4189
CAS Max % 100.0
CAS Min % 94.0
Packaging Glass bottle
Chemical Name or Material 2-Acetamidofluorene
MDL Number MFCD00001116
Molecular Formula C15H13NO
Assay Percent Range 95%
CAS 53-96-3
Infrared Spectrum Authentic
Synonym N-(2-Fluorenyl)acetamide
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause cancer.
Harmful if swallowed.
Safety and Handling

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause cancer.
Harmful if swallowed.