Please login to your online account to display your discounted pricing

2-Acetamidofluorene, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  102300050

 View more versions of this product

Catalog No. AC102300050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 223.27
Quantity 5g
Color White to yellow-brown
Beilstein 12, 1331
Merck Index 15, 4189
Melting Point 192° to 196°C
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 2-Acetamidofluorene
Assay Percent Range 95%
Molecular Formula C15H13NO
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
MDL Number MFCD00001116
CAS 53-96-3
Packaging Glass bottle
Synonym N-(2-Fluorenyl)acetamide
Health Hazard 2 GHS H Statement:
May cause cancer.
Harmful if swallowed.

Specifications

Chemical Name or Material 2-Acetamidofluorene
CAS 53-96-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C15H13NO
Formula Weight 223.27
MDL Number MFCD00001116
Synonym N-(2-Fluorenyl)acetamide
Accessories

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause cancer.
Harmful if swallowed.

SDS & Safety Documents