missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetamido-6-chlorobenzoic acid, 99%

Manufacturer:  Alfa Aesar™ L0863214

 View more versions of this product

Catalog No. AAL0863214



Specifications

Specifications

2-Acetamido-6-chlorobenzoic acid
25g
C9H8ClNO3
784450
VFHSJTHAMJFUCK-UHFFFAOYSA-N
2-acetamido-6-chlorobenzoic acid
88044
99%
216°C to 218°C
19407-42-2
MFCD00051939
2-acetylamino-6-chlorobenzoic acid, 2-n-acetylamino-6-chlorobenzoic acid, 2-chloro-6-acetamidobenzoic acid, benzoic acid, 2-acetylamino-6-chloro, acmc-20ao6f, 2-acetamido-6-chloro-benzoic acid, 2-acetylamino-6-chlorobenzoic acid, 6-chloro-2-methylcarbonylamino benzoic acid
CC(=O)NC1=C(C(=CC=C1)Cl)C(=O)O
213.617
213.62

Chemical Identifiers

19407-42-2
213.617
VFHSJTHAMJFUCK-UHFFFAOYSA-N
88044
CC(=O)NC1=C(C(=CC=C1)Cl)C(=O)O
C9H8ClNO3
MFCD00051939
2-acetylamino-6-chlorobenzoic acid, 2-n-acetylamino-6-chlorobenzoic acid, 2-chloro-6-acetamidobenzoic acid, benzoic acid, 2-acetylamino-6-chloro, acmc-20ao6f, 2-acetamido-6-chloro-benzoic acid, 2-acetylamino-6-chlorobenzoic acid, 6-chloro-2-methylcarbonylamino benzoic acid
2-acetamido-6-chlorobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents