Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetamido-6-chlorobenzoic acid, 99%

$29.69 - $328.47

Chemical Identifiers

Chemical Name or Material 2-Acetamido-6-chlorobenzoic acid
CAS 19407-42-2
Molecular Formula C9H8ClNO3
Formula Weight 213.62
MDL Number MFCD00051939
Beilstein 784450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0863203

  SDS
alfa aesar™
L0863203
1g Each for $29.69

AAL0863206

  SDS
alfa aesar™
L0863206
5g Each for $85.43

AAL0863214

  SDS
alfa aesar™
L0863214
25g Each for $328.47
Description & Specifications

Specifications

Formula Weight 213.62
Chemical Name or Material 2-Acetamido-6-chlorobenzoic acid
MDL Number MFCD00051939
CAS 19407-42-2
Molecular Formula C9H8ClNO3
Assay 99%
Beilstein 784450
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 216° to 218°C
Health Hazard 1 Warning
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.