missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetamido-5-nitrothiazole, 98%

$94.69 - $323.85

Chemical Identifiers

CAS 140-40-9
Molecular Formula C5H5N3O3S
Molecular Weight (g/mol) 187.173
MDL Number MFCD00022438
InChI Key UJRRDDHEMZLWFI-UHFFFAOYSA-N
Synonym nithiamide, 2-acetamido-5-nitrothiazole, aminitrozole, acinitrazole, nitazole, tritheon, gynofon, ametoterina, aminitrozol, lavoflagin
PubChem CID 8798
IUPAC Name N-(5-nitro-1,3-thiazol-2-yl)acetamide
SMILES CC(=O)NC1=NC=C(S1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0135603 View Documents Alfa Aesar™
L0135603
1g Each for $94.69
 
AAL0135606 View Documents Alfa Aesar™
L0135606
5g Each for $323.85
 
Specifications

Chemical Identifiers

140-40-9
187.173
UJRRDDHEMZLWFI-UHFFFAOYSA-N
8798
CC(=O)NC1=NC=C(S1)[N+](=O)[O-]
C5H5N3O3S
MFCD00022438
nithiamide, 2-acetamido-5-nitrothiazole, aminitrozole, acinitrazole, nitazole, tritheon, gynofon, ametoterina, aminitrozol, lavoflagin
N-(5-nitro-1,3-thiazol-2-yl)acetamide

Specifications

~263°C
140-40-9
MFCD00022438
14,410
UJRRDDHEMZLWFI-UHFFFAOYSA-N
N-(5-nitro-1,3-thiazol-2-yl)acetamide
8798
98%
1g
C5H5N3O3S
167361
nithiamide, 2-acetamido-5-nitrothiazole, aminitrozole, acinitrazole, nitazole, tritheon, gynofon, ametoterina, aminitrozol, lavoflagin
CC(=O)NC1=NC=C(S1)[N+](=O)[O-]
187.173
187.18
2-Acetamido-5-nitrothiazole
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XJ1570000