Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetamido-5-nitropyridine, 98%

$47.32 - $142.24

Chemical Identifiers

Chemical Name or Material 2-Acetamido-5-nitropyridine
CAS 5093-64-1
Molecular Formula C7H7N3O3
Formula Weight 181.15
MDL Number MFCD00833973
Beilstein 162973
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2000503

  SDS
alfa aesar™
L2000503
1g Each for $47.32

AAL2000506

  SDS
alfa aesar™
L2000506
5g Each for $142.24
Description & Specifications

Specifications

Formula Weight 181.15
Chemical Name or Material 2-Acetamido-5-nitropyridine
Assay 98%
Molecular Formula C7H7N3O3
MDL Number MFCD00833973
CAS 5093-64-1
Health Hazard 2 GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Beilstein 162973
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 198° to 202°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.