missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetamido-5-chlorothiazole, 97%

$42.86 - $99.47

Chemical Identifiers

CAS 20256-39-7
Molecular Formula C5H5ClN2OS
Molecular Weight (g/mol) 176.618
MDL Number MFCD01318098
InChI Key YLRXYIGXXMJPOP-UHFFFAOYSA-N
Synonym 2-acetamido-5-chlorothiazole, n-5-chlorothiazol-2-yl acetamide, n-5-chloro-1,3-thiazol-2-yl acetamide, acmc-20ao6g, 2-acetamido-5-chlorthiazol, 2-acetylamino-5-chlorothiazole, n-5-chloro-2-thiazolyl acetamide, 2-acetamido-5-chlorothiazole, acetamide,n-5-chloro-2-thiazolyl, n-5-chloranyl-1,3-thiazol-2-yl ethanamide
PubChem CID 4684134
IUPAC Name N-(5-chloro-1,3-thiazol-2-yl)acetamide
SMILES CC(=O)NC1=NC=C(S1)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1715106 View Documents Alfa Aesar™
L1715106
5g Each for $99.47
 
AAL1715103 View Documents Alfa Aesar™
L1715103
1g Each for $42.86
 
Specifications

Chemical Identifiers

20256-39-7
176.618
YLRXYIGXXMJPOP-UHFFFAOYSA-N
4684134
CC(=O)NC1=NC=C(S1)Cl
C5H5ClN2OS
MFCD01318098
2-acetamido-5-chlorothiazole, n-5-chlorothiazol-2-yl acetamide, n-5-chloro-1,3-thiazol-2-yl acetamide, acmc-20ao6g, 2-acetamido-5-chlorthiazol, 2-acetylamino-5-chlorothiazole, n-5-chloro-2-thiazolyl acetamide, 2-acetamido-5-chlorothiazole, acetamide,n-5-chloro-2-thiazolyl, n-5-chloranyl-1,3-thiazol-2-yl ethanamide
N-(5-chloro-1,3-thiazol-2-yl)acetamide

Specifications

208°C to 212°C
20256-39-7
MFCD01318098
2-acetamido-5-chlorothiazole, n-5-chlorothiazol-2-yl acetamide, n-5-chloro-1,3-thiazol-2-yl acetamide, acmc-20ao6g, 2-acetamido-5-chlorthiazol, 2-acetylamino-5-chlorothiazole, n-5-chloro-2-thiazolyl acetamide, 2-acetamido-5-chlorothiazole, acetamide,n-5-chloro-2-thiazolyl, n-5-chloranyl-1,3-thiazol-2-yl ethanamide
CC(=O)NC1=NC=C(S1)Cl
176.618
176.63
2-Acetamido-5-chlorothiazole
5g
C5H5ClN2OS
120475
YLRXYIGXXMJPOP-UHFFFAOYSA-N
N-(5-chloro-1,3-thiazol-2-yl)acetamide
4684134
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning