missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Acetamido-5-bromopyridine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442420010

 View more versions of this product

Catalog No. AC442420010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Acetamido-5-bromopyridine
176°C to 178°C
7169-97-3
C7H7BrN2O
MJFCOXATGBYERZ-UHFFFAOYSA-N
N-(5-bromopyridin-2-yl)acetamide
293097
98%
Glass Bottle
1g
97.5% min. (HPLC)
2-acetamido-5-bromopyridine, 2-acetylamino-5-bromopyridine, acetamide, n-5-bromo-2-pyridinyl, n-5-bromo-2-pyridinyl acetamide, n-5-bromo-2-pyridyl acetamide, n-5-bromo-pyridin-2-yl-acetamide, n-5-bromopyridin-2-yl acetamide, n-acetylamino-5-bromopyridine, pubchem5546
CC(=O)NC1=NC=C(C=C1)Br
215.05
215.05

Chemical Identifiers

7169-97-3
215.05
2-acetamido-5-bromopyridine, 2-acetylamino-5-bromopyridine, acetamide, n-5-bromo-2-pyridinyl, n-5-bromo-2-pyridinyl acetamide, n-5-bromo-2-pyridyl acetamide, n-5-bromo-pyridin-2-yl-acetamide, n-5-bromopyridin-2-yl acetamide, n-acetylamino-5-bromopyridine, pubchem5546
N-(5-bromopyridin-2-yl)acetamide
C7H7BrN2O
MJFCOXATGBYERZ-UHFFFAOYSA-N
293097
CC(=O)NC1=NC=C(C=C1)Br
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents