Please login to your online account to display your discounted pricing

2-Acetamido-5-bromopyridine, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Acetamido-5-bromopyridine
Assay Percent Range 98%
Molecular Formula C7H7BrN2O
Formula Weight 215.05
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC442420050

  SDS
thermo fisher scientific
AC442420050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC442420010

  SDS
thermo fisher scientific
AC442420010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Acetamido-5-bromopyridine
CAS 7169-97-3
Assay Percent Range 98%
Molecular Formula C7H7BrN2O
Formula Weight 215.05
Quantity 5g