Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetamido-5-bromopyridine, 98%

$137.26 - $137.26

Chemical Identifiers

Chemical Name or Material 2-Acetamido-5-bromopyridine
CAS 7169-97-3
Molecular Formula C7H7BrN2O
Formula Weight 215.05
MDL Number MFCD00468968
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2745609

  SDS
alfa aesar™
H2745609
10g Each for $137.26
Description & Specifications

Specifications

Formula Weight 215.05
Chemical Name or Material 2-Acetamido-5-bromopyridine
Assay 98%
Molecular Formula C7H7BrN2O
MDL Number MFCD00468968
CAS 7169-97-3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 175° to 179°C
Quantity 10g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.