Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetamido-5-bromobenzoic acid, 98%

$42.64 - $140.88

Chemical Identifiers

Chemical Name or Material 2-Acetamido-5-bromobenzoic acid
CAS 38985-79-4
Molecular Formula C9H8BrNO3
Formula Weight 258.07
MDL Number MFCD00040904
Beilstein 882463
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0213906

  SDS
alfa aesar™
L0213906
5g Each for $42.64

AAL0213914

  SDS
alfa aesar™
L0213914
25g Each for $140.88
Description & Specifications

Specifications

Formula Weight 258.07
Chemical Name or Material 2-Acetamido-5-bromobenzoic acid
Molecular Formula C9H8BrNO3
CAS 38985-79-4
MDL Number MFCD00040904
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Melting Point 222° to 224°C
Beilstein 882463
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.