Please login to your online account to display your discounted pricing

2-Acetamido-5-bromobenzoic acid, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  35402-0050

 View more versions of this product

Catalog No. AC354020050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystals or Crystalline Powder
Formula Weight 258.07
Quantity 5g
Color Pale brown
TSCA TSCA
Melting Point 218° to 223°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-Acetamido-5-bromobenzoic acid
Assay Percent Range 97%
Molecular Formula C9H8BrNO3
Linear Formula CH3C(o)nHC6H3(Br)COOH
MDL Number MFCD00040904
CAS 38985-79-4
Name Note 97%
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 2-Acetamido-5-bromobenzoic acid
Name Note 97%
CAS 38985-79-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H8BrNO3
Linear Formula CH3C(o)nHC6H3(Br)COOH
Formula Weight 258.07
MDL Number MFCD00040904