missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
2-Acetamido-5-bromobenzoic acid, 97%, ACROS Organics™
Chemical Identifiers
CAS |
38985-79-4 |
Molecular Formula |
C9H8BrNO3 |
Molecular Weight (g/mol) |
258.07 |
MDL Number |
MFCD00040904 |
InChI Key |
QVABAFHRLMDDLM-UHFFFAOYSA-N |
Synonym |
2-acetamido-5-bromobenzoic acid, 2-acetamino-5-bromobenzoic acid, 2-acetylamino-5-bromobenzoic acid, 5-bromo-2-acetamidobenzoic acid, benzoic acid, 2-acetylamino-5-bromo, benzoic acid,2-acetylamino-5-bromo, n-acetyl-5-bromo-anthranilic acid, n-acetyl-5-bromoanthranilic acid |
PubChem CID |
520935 |
IUPAC Name |
2-acetamido-5-bromobenzoic acid |
SMILES |
CC(=O)NC1=C(C=C(C=C1)Br)C(=O)O |
View More Specs
Products
1
Description
Specifications
Chemical Identifiers
|
38985-79-4 |
|
258.07 |
|
QVABAFHRLMDDLM-UHFFFAOYSA-N |
|
520935 |
|
CC(=O)NC1=C(C=C(C=C1)Br)C(=O)O |
|
C9H8BrNO3 |
|
MFCD00040904 |
|
2-acetamido-5-bromobenzoic acid, 2-acetamino-5-bromobenzoic acid, 2-acetylamino-5-bromobenzoic acid, 5-bromo-2-acetamidobenzoic acid, benzoic acid, 2-acetylamino-5-bromo, benzoic acid,2-acetylamino-5-bromo, n-acetyl-5-bromo-anthranilic acid, n-acetyl-5-bromoanthranilic acid |
|
2-acetamido-5-bromobenzoic acid |
Specifications
|
Authentic |
|
Brown |
|
5g |
|
96.0 |
|
96% min. (HPLC) |
|
CH3C(o)NHC6H3(Br)COOH |
|
2-acetamido-5-bromobenzoic acid, 2-acetamino-5-bromobenzoic acid, 2-acetylamino-5-bromobenzoic acid, 5-bromo-2-acetamidobenzoic acid, benzoic acid, 2-acetylamino-5-bromo, benzoic acid,2-acetylamino-5-bromo, n-acetyl-5-bromo-anthranilic acid, n-acetyl-5-bromoanthranilic acid |
|
CC(=O)NC1=C(C=C(C=C1)Br)C(=O)O |
|
258.07 |
|
258.07 |
|
97% |
|
Glass bottle |
|
218°C to 223°C |
|
38985-79-4 |
|
100.0 |
|
C9H8BrNO3 |
|
MFCD00040904 |
|
QVABAFHRLMDDLM-UHFFFAOYSA-N |
|
2-acetamido-5-bromobenzoic acid |
|
520935 |
|
Crystals or Crystalline Powder |
|
2-Acetamido-5-bromobenzoic acid, 97% |
Safety and Handling
SDS