Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetamido-5-aminopyridine, 98%

$59.72 - $184.72

Chemical Identifiers

Chemical Name or Material 2-Acetamido-5-aminopyridine
CAS 29958-14-3
Molecular Formula C7H9N3O
Formula Weight 151.17
MDL Number MFCD00160311
Beilstein 128346
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2000403

 
alfa aesar™
L2000403
1g Each for $59.72

AAL2000406

 
alfa aesar™
L2000406
5g Each for $184.72
Description & Specifications

Specifications

Formula Weight 151.17
Chemical Name or Material 2-Acetamido-5-aminopyridine
Assay 98%
Molecular Formula C7H9N3O
MDL Number MFCD00160311
CAS 29958-14-3
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 128346
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 144° to 154°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.