missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetamido-4-(4-bromophenyl)thiazole, 97%

$43.64 - $137.88

Chemical Identifiers

CAS 53173-91-4
Molecular Formula C11H9BrN2OS
Molecular Weight (g/mol) 297.17
InChI Key CALREGGMKYUGIP-UHFFFAOYSA-N
Synonym n-4-4-bromophenyl-1,3-thiazol-2-yl acetamide, 2-acetamido-4-4-bromophenyl thiazole, 2-n-acetamino-4-4-bromophenyl thiazole, n-4-4-bromophenyl thiazol-2-yl acetamide
PubChem CID 790490
IUPAC Name N-[4-(4-bromophenyl)-1,3-thiazol-2-yl]acetamide
SMILES CC(=O)NC1=NC(=CS1)C2=CC=C(C=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66025MD View Documents Alfa Aesar™
H66025MD
250mg Each for $43.64
 
AAH6602503 View Documents Alfa Aesar™
H6602503
1g Each for $137.88
 
Specifications

Chemical Identifiers

53173-91-4
297.17
n-4-4-bromophenyl-1,3-thiazol-2-yl acetamide, 2-acetamido-4-4-bromophenyl thiazole, 2-n-acetamino-4-4-bromophenyl thiazole, n-4-4-bromophenyl thiazol-2-yl acetamide
N-[4-(4-bromophenyl)-1,3-thiazol-2-yl]acetamide
C11H9BrN2OS
CALREGGMKYUGIP-UHFFFAOYSA-N
790490
CC(=O)NC1=NC(=CS1)C2=CC=C(C=C2)Br

Specifications

White to Yellow
250mg
C11H9BrN2OS
CALREGGMKYUGIP-UHFFFAOYSA-N
N-[4-(4-bromophenyl)-1,3-thiazol-2-yl]acetamide
790490
Powder
2-Acetamido-4-(4-bromophenyl)thiazole
233°C to 234°C
53173-91-4
n-4-4-bromophenyl-1,3-thiazol-2-yl acetamide, 2-acetamido-4-4-bromophenyl thiazole, 2-n-acetamino-4-4-bromophenyl thiazole, n-4-4-bromophenyl thiazol-2-yl acetamide
CC(=O)NC1=NC(=CS1)C2=CC=C(C=C2)Br
297.17
297.17
97%